Mouse recombinant CD40L (soluble)
Catalog # ENZOALX5220702010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Mouse
- Seq:02
- Biological activity:Stimulates the proliferation of mouse B cells and dendritic cells. The activity of rmsCD40L increases 1'000-fold (stimulation in the ng/ml ra nge) in the presence of a cross-linki ng enhancer (see Set Prod. No. ALX-850-075).
- Reconstitution instructions:Reconstitute each tube with 100 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
- Protein/peptide name:CD40L (soluble)
- Purity:>90% (SDS-PAGE)
- Concentration:0,1 mg/ml after reconstitution
- Formulation:Lyophilized. Contains PBS.
Specifications
About this item
Produced in HEK 293 cells. The extracellular domain of mouse CD40L (CD154) (aa 115-260) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG®-tag.