Human recombinant TWEAK (soluble) (from E. coli)
Catalog # ENZOALX522021C010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Biological activity:Kills TWEAK sensitive cells. Has a pro-apoptotic effect on several target cell lines. Induces angiogenesis and proliferation in endothelial cells.
- Reconstitution instructions:Reconstitute with 100 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum.
- Protein/peptide name:TWEAK (soluble)
- Purity:>95% (SDS-PAGE)
- Concentration:0,1 mg/ml after reconstitution
- Formulation:Lyophilised. Contains PBS.
Specifications
About this item
Produced in E. coli. The extracellular domain of human TWEAK (aa 106-249) is fused at the N-terminus to a linker peptide (11 aa) and a FLAG®-tag.