Human recombinant CD40L (soluble) (from E. coli)
Supplier: Enzo Life Sciences
Certificates
About this item
Produced in E. coli. The extracellular domain of human CD40L (CD154) (aa 116-261) is fused at the N-terminus to a linker peptide (6 aa) and a FLAG®-tag.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Biological activity:Stimulates growth of B cells. The activity of rhsCD40L increases 1'000-fold (stimulation in the ng/ml ra nge) in the presence of cross-linki ng enhancer (see Set Prod. No. ALX-850-064). For stimulation of mouse cells via CD40 use CD40L, Soluble (mouse) (rec
- Protein/peptide name:CD40L (soluble)
- Purity:>95% (SDS-PAGE)
- Concentration:0,1 mg/ml after reconstitution
- Formulation:Lyophilized. Contains PBS.
Specifications
Frequently Bought Together