Mouse nerve growth factor 7S (from submaxillary glands)
Catalog # ENZOALX521008C100
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:100 µG
- Source:Submaxillary glands
- Species:Mouse
- Seq:02
- Storage conditions:−20 °C
- Biological activity:Bioassayed for neurotrophic activity using the rat pheochromocytoma cell PC12 cultures over a period of 7 - 14 days. Neurotrophic activity is in the range of 0,5 - 50 ng/ml. Recommended concentration to be used in vitro for maintenance of sympathetic and immature sensory nerve cells is 50 - 100 ng/ml medium.
- Reconstitution instructions:Dilute in sterile water or DMEM to a concentration of at least 10 µg/ ml and store at –80 °C before use. Medium containing NGF cannot be reused due to NGF degradation.
- Protein synonyms:NGF 7S
- Protein/peptide name:Nerve Growth Factor 7S
- Purity:>97% (SDS-PAGE)
- Molecular weight:~130kDa
- Formulation:Lyophilised
- Shipping temperature:Shipped on blue ice
Specifications
About this item
Isolated from mouse submaxillary glands by a modification of the procedure described by T. Darling, et al.; Cell Culture Meth. Mol. Cell Biol. 4, 79 (1984).
Quantity: Determined by Lowry and Pierce method.