Human recombinant SUMO2 (1-93) (from E. coli)
Catalog # ENZOALX201089C500
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:500 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:SUMO2 (1-93)
- Purity:>95%
- Formulation:Lyophilised
Specifications
About this item
Active form of SUMO-2
Produced in E. coli.
Active form of human SUMO-2 for conjugation to protein substrates.