- Pk:5 µG
- Source:Fibroblasts
- Species:Human
- Seq:02
- Environmentally Preferable:
- Protein/peptide name:MMP-1 proenzyme
- Purity:>90% (SDS-PAGE, Western blot)
- Concentration:~80 µg/ml (Pierce-BCA)
- Formulation:Liquid. In 50 mM TRIS-HCl, pH 7,0, 300 mM NaCl, 5 mM CaCl₂, 1 µM ZnCl₂, 0,05% BRIJ35 and 0,05% sodium azide.
- Purification:No other MMP contaminants are detectable
Isolated from human rheumatoid synovial fibroblasts. Requires activation.