Order Entry
Netherlands
ContactUsLinkComponent
ETA receptor antagonist [BQ-610]
  ENZOALX155011P001
 :  
ETA receptor antagonist [BQ-610]
  ENZOALX155011P001
 :  
  • Pk:
    1 mg
  • Seq:
    02
  • Protein/peptide name:
    ETA receptor antagonist [BQ-610]
30-fold more potent ETA antagonist than BQ-123 in pig aortic smooth muscle membrane assay, same activity as BQ-123 when tested on pig cerebellum membrane.
 : Quantity: *Sold as net peptide weight.