N-Acetyl-Asp-Glu-OH
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Glutamate receptor ligand
Endogenous neurotransmitter with high affinity for the brain glutamate receptor. N-acetylated α-linked acidic dipeptidase (NAALADase), a membrane-bound peptidase, hydrolyzes α-NAAG, a major brain peptide, to N-acetylaspartate and glutamate. Discriminates between mGluR2 and mGluR3. mGluR3 activator. NMDA receptor antagonist. Neuroprotective.
Specifications
- Protein/peptide name:N-Acetyl-Asp-Glu-OH
- Purity:>98%