Human recombinant Erythropoietin alpha (from CHO cells)
Catalog # ENZOADI908050500
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 Vial
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Seq:02
- Protein/peptide name:Erythropoietin alpha
- Purity:>98% (SDS-PAGE; RP-HPLC)
- Formulation:Lyophilized. Each milligram contains 5,8 mg sodium chloride and 0,06 mg citric acid. Reconstitute with sterile 18W - cm water not <100 μg/ml
- Purification:Purified by multi-step chromatography.
Specifications
About this item
Produced in CHO cells.