Order Entry
Netherlands
ContactUsLinkComponent
Calcium assay kit, FluoForte®
Calcium assay kit, FluoForte®
Supplier:  ENZO LIFE SCIENCES
Certificates
undefined
Calcium assay kit, FluoForte®
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Storage conditions:
    –80 °C (long term), –20 °C (short term)
  • Application:
    Flow cytometry, Fluorescence microscopy, Fluorescent detection, HTS

Specifications

Frequently Bought Together