Order Entry
Netherlands
ContactUsLinkComponent
Anti-OPN Mouse Monoclonal Antibody [clone: 53]
Anti-OPN Mouse Monoclonal Antibody [clone: 53]
Catalog # ENZOADI905629100
Supplier:  ENZO LIFE SCIENCES
Anti-OPN Mouse Monoclonal Antibody [clone: 53]
Catalog # ENZOADI905629100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Osteopontin
  • Antigen symbol:
    OPN
  • Clonality:
    Monoclonal
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS containing 0,1% sodium azide.
  • Antigen synonyms:
    early T-lymphocyte activation 1|bone sialoprotein I|osteopontin|SPP-1|nephropontin|secreted phosphoprotein 1 (osteopontin|bone sialoprotein 1|secreted phosphoprotein-1 (osteopontin|bone sialoprotein)|urinary stone protein|uropontin|SPP1/CALPHA1 fusion|osteopontin/immunoglobulin alpha 1 heavy chain constant region fusion protein|early T-lymphocyte activation 1)
  • Purification:
    Protein G affinity purified
  • Pk:
    100 µG

Specifications

About this item

Osteopontin (OPN) is an acidic extracellular matrix cell adhesion protein that is relatively abundant not only in bone matrix, plasma, urine, and milk, but is also found in malignant and atherogenic tissues. Phosphorylation, glycosylation and calcium modifications allow intact and fragmented Osteopontin to direct a variety of diverse responses including tissue remodeling, inflammation and cell survival. Plasma Osteopontin has been shown to provide prognostic information in breast, prostate, colon, and lung cancers as well as metastatic carcinomas. The notable presence of Osteopontin in a variety of tumors is strongly correlated to pathological stage, suggesting its critical role in tumor invasiveness, progression and metastasis. In addition, Osteopontin inhibits inducible nitric oxide synthase activity, thereby protecting tumor cells from NO-mediated macrophage cytotoxic attack. Osteopontin is found in atherosclerotic plaques and may drive a number of diabetic vascular pathologies.

Type: Primary
Antigen: OPN
Clonality: Monoclonal
Clone: 53
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human