Order Entry
Netherlands
ContactUsLinkComponent
Anti-Methyl-lysine Rabbit Polyclonal Antibody
Anti-Methyl-lysine Rabbit Polyclonal Antibody
Catalog # ENZOADIKAPTF121E
Supplier:  ENZO LIFE SCIENCES
Anti-Methyl-lysine Rabbit Polyclonal Antibody
Catalog # ENZOADIKAPTF121E
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Methyl-Lysine
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid
  • Storage buffer:
    PBS containing 50% glycerol
  • Storage temperature:
    −20 °C
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Methylated KLH
  • Purification:
    Purified by Immunoaffinity chromatography
  • Pk:
    100 µG

Specifications

About this item

Acetylation and methylation of lysine are important post-translational modifications that regulate numerous protein-protein and protein-DNA interactions. Lysine acetylation and methylation involves the transfer of acetylCoA, or one or more methyl groups, to the e-amino group of lysine by modifying enzymes and cofactors. Histones and transcription factors are the primary targets of lysine acetylation and methylation, with either modification capable of inducing gene silencing or expression due to differential regulation of cofactors. For example, varying degrees of mono-, di-, and tri-methylation or acetylation of histone H3 at lysine residue 9 are known to demark distinct chromatin regions during various states of gene activation (methylation) or repression (acetylation).

Type: Primary
Antigen: Methyl-lysine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Species independent