Anti-Pyruvate dehydrogenase Kinase 1 Rabbit Polyclonal Antibody
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
PDK-1 (Pyruvate dehydrogenase kinase-1) is a 48 kDa protein kinase. It is a member of the mitrochondrial matrix protein kinase family and is related to the histidine protein kinases found in prokaryotes. PDK-1 is responsible for phosphorylation and concomitant inactivation of pyruvate dehydrogenase.
Type: Primary
Antigen: Pyruvate dehydrogenase kinase 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Rabbit, Pig, Monkey, Dog
Specifications
- Catalog No:
- ENZOADIKAPPK112F
- ENZOADIKAPPK112D
- Antigen symbol:
- PDK1
- PDK1
- Antigen name:
- Pyruvate dehydrogenase Kinase 1
- Pyruvate dehydrogenase Kinase 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Pig, Rabbit, Monkey, Human, Dog, Rat, Mouse
- Pig, Rabbit, Monkey, Human, Dog, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide corresponding to a portion of human PDK1 (Pyruvate dehydrogenase kinase-1)
- Synthetic peptide corresponding to a portion of human PDK1 (Pyruvate dehydrogenase kinase-1)
- Form:
- Liquid
- Liquid
- Purification:
- Purified by Protein A affinity chromatography
- Storage buffer:
- PBS containing 50% glycerol and 0.09% sodium azide
- PBS containing 50% glycerol and 0.09% sodium azide
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together