Anti-EIF2 alpha Rabbit Polyclonal Antibody
Type: Primary
Antigen: EIF2 alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Sheep, Rabbit, Pig, Monkey, Hamster, Guinea pig, Dog, Bovine
- Catalog No:
- ENZOADIKAPCP130F
- ENZOADIKAPCP130D
- Antigen symbol:
- EIF2 alpha
- EIF2 alpha
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Hamster, Pig, Rabbit, Monkey, Sheep, Human, Bovine, Guinea pig, Dog, Rat, Mouse
- Hamster, Pig, Rabbit, Monkey, Sheep, Human, Bovine, Guinea pig, Dog, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide corresponding to a portion of rat eIF2-alpha
- Synthetic peptide corresponding to a portion of rat eIF2-alpha
- Form:
- Liquid
- Liquid
- Purification:
- Purified by Immunoaffinity chromatography
- Purified by Immunoaffinity chromatography
- Storage buffer:
- PBS, pH 7,2, containing 0,09% sodium azide and 50% glycerol.
- PBS, pH 7,2, containing 0,09% sodium azide and 50% glycerol.
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together