Anti-MHC1 Mouse Monoclonal Antibody
Catalog # ENZOALX805711C100
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen name:MHC Class I
- Antigen symbol:MHC1
- Clonality:Monoclonal
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Western blot:Yes
- Seq:02
- Form:Liquid. In PBS, pH 7,4. Sterile filtered. Contains no preservatives
- Concentration:1 mg/ml
- Purification:≥95% (SDS-PAGE)
- Size:100 µg
- Pk:100 µG
Specifications
About this item
HLA-class I major histocompatibility (MHC) antigens are intrinsic membrane glycoproteins expressed on nucleated cells and noncovalently associated with an invariant beta2 microglobulin. They carry foreign determinants important for immune recognition by cytotoxic T cells, thus important for anti-viral and anti-tumour defence. Human HLA-class I antigens are represented by HLA-A, HLA-B and HLA-C molecules.
- For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Recognizes human, monkey, bovine and cat MHC class I. Does not cross-react with rabbit MHC class I. Recognizes virtually all nucleated human cells. Suitable as positive control for HLA tissue typing and crossmatching.
Type: Primary
Antigen: MHC1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human