Anti-BIMS/EL/L Rat Monoclonal Antibody
Catalog # ENZOALX804528C100
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen symbol:BIMS/EL/L
- Clonality:Monoclonal
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Western blot:Yes
- Seq:02
- Form:Liquid. Protein G affinity purified antibody in PBS containing 25% glycerol and 0,02% sodium azide
- Concentration:1 mg/ml
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Host: Rat, Isotype: IgG2a
- For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Recognizes an epitope within aa 20-40 of human, mouse, rat, monkey and dog BimL, BimEL, and BimS. Also recognizes the six novel isoforms of human Bim, generated by alternative splicing and lacking the C-terminal hydrophobic region recently identified.
Type: Primary
Antigen: BIMS/EL/L
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human