Order Entry
Netherlands
ContactUsLinkComponent
Anti-BIMS/EL/L Rat Monoclonal Antibody
Anti-BIMS/EL/L Rat Monoclonal Antibody
  ENZOALX804527C100
Anti-BIMS/EL/L Rat Monoclonal Antibody
  ENZOALX804527C100

 

  • Antibody type:
    Primary
  • Antigen symbol:
    BIMS/EL/L
  • Clonality:
    Monoclonal
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS containing 0,02% sodium azide and 25% glycerol
  • Concentration:
    1 mg/ml
  • Purification:
    Protein G-affinity purified
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Host: Rat, Isotype: IgG2a

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes an epitope within aa 20-40 of human, mouse, rat, monkey and dog BimL (~19kDa), BimEL (~23kDa), and BimS (~16kDa). Also recognizes the six novel isoforms of human Bim, generated by alternative splicing and lacking the C-terminal hydrophobic region recently identified.

Type: Primary
Antigen: BIMS/EL/L
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human