Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-CD14 Mouse Monoclonal Antibody
Catalogus # ENZOALX804498C100
Supplier:  ENZO LIFE SCIENCES
Anti-CD14 Mouse Monoclonal Antibody
Catalogus # ENZOALX804498C100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD14 Molecule
  • Antigen symbol:
    CD14
  • Clonality:
    Monoclonal
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Seq:
    02
  • Form:
    Lyophilised from PBS. Contains no preservatives
  • Antigen synonyms:
    monocyte differentiation antigen CD14|myeloid cell-specific leucine-rich glycoprotein
  • Concentration:
    1 mg/ml after reconstitution.
  • Purification:
    Protein G-affinity purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

The CD14 glycoprotein, gp 55, is present on most monocytic and macrophage like cell types: monocytes, macrophages, Kupffer cells, pleural phagocytic cells and dendritic reticular cells. CD14 is also observed on granulocytes and activated or transformed B-cells. Furthermore CD14 is present in a soluble form in human serum, urine and other body fluids. The CD14 molecule has been reported to be a receptor for endotoxin.

  • For ELISA, Flow Cytometry, Immunoprecipitation

Recognizes human (epitope aa 147-152) CD14.

Reconstitute with 100µl distilled water.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human