Order Entry
Netherlands
ContactUsLinkComponent
Anti-Enhancer for Ligands Monoclonal Antibody
Anti-Enhancer for Ligands Monoclonal Antibody
Catalogus # ENZOALX804034C050
Supplier:  ENZO LIFE SCIENCES
undefined
Anti-Enhancer for Ligands Monoclonal Antibody
Catalogus # ENZOALX804034C050
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Enhancer for ligands
  • Clonality:
    Monoclonal
  • Seq:
    02
  • Form:
    Liquid
  • Storage buffer:
    PBS, pH 7,4, containing 50% glycerol. The formulation contains no antimicrobial preservatives.
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: Enhancer for Ligands
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: