Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-MRP1 Mouse Monoclonal Antibody
 
Anti-MRP1 Mouse Monoclonal Antibody

 

  • Catalog No:
  • ENZOALX801013C250
  • ENZOALX801013C500
  • Antigen symbol:
  • MRP1
  • MRP1
  • Antigen name:
  • Multidrug resistance-associated protein 1
  • Multidrug resistance-associated protein 1
  • Antigen synonyms:
  • ABC29|ATP-binding cassette transporter variant ABCC1delta-ex13|ATP-binding cassette transporter variant ABCC1delta-ex25&,26|MRP1|LTC4 transporter|ABCC|GS-X|ATP-binding cassette transporter variant ABCC1delta-ex13&,14|leukotriene C(4) transporter|MRP|multidrug resistance-associated protein 1|ATP-binding cassette transporter variant ABCC1delta-ex25
  • ABC29|ATP-binding cassette transporter variant ABCC1delta-ex13|ATP-binding cassette transporter variant ABCC1delta-ex25&,26|MRP1|LTC4 transporter|ABCC|GS-X|ATP-binding cassette transporter variant ABCC1delta-ex13&,14|leukotriene C(4) transporter|MRP|multidrug resistance-associated protein 1|ATP-binding cassette transporter variant ABCC1delta-ex25
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Reactivity:
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Form:
  • Liquid. In serum free culture supernatant containing 0,7% BSA and 0,1% sodium azide.
  • Liquid. In serum free culture supernatant containing 0,7% BSA and 0,1% sodium azide.
  • Concentration:
  • ~500 µg/ml
  • ~500 µg/ml

 

Frequently Bought Together