Anti-ENOS Rabbit Polyclonal Antibody
Catalog # ENZOALX210509R100
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen name:Endothelial Nitric Oxide Synthase
- Antigen symbol:ENOS
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Form:Liquid. In PBS containing 1 mg/ml BSA and 0,05% sodium azide.
- Antigen synonyms:eNOS|nitric oxide synthase 3 endothelial cell|ECNOS|NOS3
- Concentration:0,2 mg/ml
- Purification:Epitope-affinity purified IgG
- Pk:100 µl
Specifications
About this item
Host: Rabbit
- For Immunohistochemistry, Western Blot
Recognizes human, mouse, rat, bovine, pig and dog eNOS (NOS III). Detects a band of ~140kDa by Western blot. Does not cross-react with human iNOS (NOS II) or rat nNOS (NOS I).
Type: Primary
Antigen: ENOS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human