Anti-CNR1 Rabbit Polyclonal Antibody
ENZOALX210316R100
- Antibody type:Primary
- Antigen name:Cannabinoid Receptor 1
- Antigen symbol:CNR1
- Clonality:Polyclonal
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Western blot:Yes
- Seq:02
- Form:Liquid. In PBS containing 50% glycerol, 1 mg/ml BSA and 0,05% sodium azide.
- Antigen synonyms:CANN6|cannabinoid receptor 1 (brain)|CNR1|OTTHUMP00000016839|RP1-23D17.1|CB1A|CNR|CB-R|CB1K5|HGNC:2159|CB1|cannabinoid receptor 1 OTTHUMP00000016838|central cannabinoid receptor|CB1R
- Concentration:3,3 mg/ml
- Purification:Affinity purified IgG
- Pk:100 µl
Host: Rabbit
- For Immunocytochemistry, Immunohistochemistry, Western Blot
Recognizes human, mouse, rat, monkey, amphibian, chicken and fish CB1. Detects a band of ~60kDa by Western blot representing CB1; also detects less intense bands of ~23kDa, ~72kDa and ~180kDa from rat brain homogenate.
Type: Primary
Antigen: CNR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: