Specifications
- Antibody type:Primary
- Antigen name:Glutathione
- Antigen symbol:GSH
- Clonality:Monoclonal
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Western blot:Yes
- Seq:02
- Form:Liquid. In PBS, pH 7,2, containing 0,01% sodium azide.
- Purification:Protein A affinity purified
- Pk:100 µG
Specifications
About this item
Glutathione, a major intracellular non-protein thiol, protects against free radical damage by providing reducing equivalents for several key antioxidant enzymes. In addition, glutathione acts as a scavenger of hydroxyl radicals and nascent oxygen. Elevated glutathione levels are associated with a reduced rate of illness, higher levels of self-rated health, lower cholesterol, lower body mass index, and lower blood pressure among the elderly. Glutathione provides a primary defense system for the removal of oxidants in the brain. Studies have revealed a correlation between low glutathione levels and damage to neurons that manufacture dopamine, suggesting a link to Parkinson's disease.
- For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Species independent.
Type: Primary
Antigen: GLUTATHIONE
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Species Independent