Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-CASP9 Rabbit Polyclonal Antibody
  ENZOADIAAP149E
Anti-CASP9 Rabbit Polyclonal Antibody
  ENZOADIAAP149E

 

  • Antibody type:
    Primary
  • Antigen name:
    Caspase 9
  • Antigen symbol:
    CASP9
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Seq:
    02
  • Antigen synonyms:
    EC 3.4.22.62|ICE-like apoptotic protease 6,ICE-LAP6,Apoptotic protease Mch-6,Apoptotic protease-activating factor 3|Apaf-3|CASP-9
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

The caspases are a family of cysteine proteases that cleave after certain aspartate residues, and are primarily recognized as mediators of apoptosis. caspases are synthesized as inactive zymogens that can be cleaved to form active enzymes following the induction of apoptosis by stress or death receptors. Initiator caspases (e.g. caspase-8 and -10) are activated by dimerization of the zymogen on a dedicated adaptor protein. These activated initiator caspases in-turn cleave downstream effector or executioner caspases (e.g. caspase-3, -6, and -7) in a cascade-like manner, which cleave key cellular proteins that lead to the morphological changes associated with apoptotic cell death.

  • For Western Blot

Recognizes human, mouse, rat, bovine, dog, chicken, guinea pig, monkey and sheep caspase-9. Detects bands of ~46-50kDa (proform) and ~35/37kDa, ~18kDa, ~10kDa (processed) by Western blot.

Type: Primary
Antigen: CASP9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human