Anti-OPG Mouse Monoclonal Antibody
Catalog # ENZOADIAAM020E
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen name:Osteoprotegerin
- Antigen symbol:OPG
- Clonality:Monoclonal
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Seq:02
- Form:Liquid. In PBS containing 0.2% gelatin and 0.05% sodium azide.
- Antigen synonyms:osteoprotegerin|MGC29565|osteoclastogenesis inhibitory factor|OCIF|TR1|tumor necrosis factor receptor superfamily member 11B|OPG
- Purification:Protein G affinity purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Osteoprotegerin (OPG) is a secreted glycoprotein belonging to the TNF receptor (TNF-R) superfamily that functions as a decoy receptor for RANK-Ligand (RANKL/OPGL). Unlike other TNF-Rs, OPG lacks a membrane-anchoring hydrophobic domain and lacks any apparent cell-associated signaling capability. OPG secreted by osteoblasts competes with RANK (expressed on osteoblast precursors) for binding to RANKL, blocking osteoclastogenesis and increasing bone density.
- For Western Blot
Recognizes human OPG. Detects a band of ~55kDa & ~110kDa by Western blot.
Type: Primary
Antigen: OPG
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human