Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-PAGA Mouse Monoclonal Antibody
  ENZOADI905619100
Anti-PAGA Mouse Monoclonal Antibody
  ENZOADI905619100

 

  • Antibody type:
    Primary
  • Antigen name:
    Anthrax protective Antigen
  • Antigen symbol:
    PAGA
  • Clonality:
    Monoclonal
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Bacteria
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS containing 0,01% sodium azide. Concentration 1 mg/ml.
  • Antigen synonyms:
    AnthraxPA
  • Purification:
    Protein G affinity purified
  • Pk:
    100 µG

 

 

The exotoxin produced by B. anthracis is composed of three proteins: a cell-receptor binding protein known as Protective Antigen (PA), and two enzymes, Edema Factor (EF) and Lethal Factor (LF). An 83 kDa form of PA binds to a cell surface receptor and, upon proteolytic activation to a 63 kDa fragment, heptamerizes to form a membrane channel that mediates the entry of three molecules of LF or EF into the cells. PA and LF form Lethal Toxin, which is the dominant virulence factor produced by B. Anthracis and is the major cause of death in infected animals.

  • For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot

Recognizes bacteria Anthrax Protective Antigen (PA).

Type: Primary
Antigen: PAGA
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bacteria