Single channel pipettes, mechanical, fixed / variable volume, Finnpipette™ F3
Supplier: Thermo Fisher Scientific
Certificates
About this item
Superior comfort, performance, reliability and easiness of maintenance in one of the lightest pipettes available in the market.
- Advanced Volume Gearing mechanism (AVG) - Self supportive modular mechanism increasing accuracy, precision and durability, with light force and smaller increments for volume adjustment
- Large display with white numbers on black background for increased volume visibility and security
- Double action pipetting button with rotating upper part preventing accidental volume alteration
- Superior handle ergonomics and very low force pipetting movement
- Super blow-out piston for volumes of 50 µl and below, which ensures the delivery of micro-size drops
- Autoclavable (tip cone and tip ejector)
* Micro
Ordering information: Supplied with service tool, Finntip sample, tube of grease, shelf hanger, two stickers, calibration certificate and instruction manual.
Specifications
- Catalog No:
- LABS4640100
- 613-1473
- 613-1464
- 613-1472
- 613-1483
- LABS4650090
- 613-1482
- 613-1466
- 613-1475
- 613-1474
- 613-1465
- LABS4650100
- 613-1470
- 613-1461
- 613-1469
- 613-1460
- 613-1468
- 613-1459
- 613-1467
- 613-1463
- 613-1471
- 613-1462
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS
- ISO14001 (Certificate Registration No. EMS 623400)
- IRECS
- ISO 14001 (Certificate Registration No. EMS 623400)
- IRECS
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- IRECS
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- IRECS
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- IRECS
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- IRECS
- ISO 14001 (Certificate Registration No. EMS 623400)
- IRECS
- ISO 14001 (Certificate Registration No. EMS 623400)
- IRECS
Specifications
Starter Pack
Product Information
Frequently Bought Together