Organic reference standard, 5-chloro-2-methyl-4-isothiazolin-3-one, Dr. Ehrenstorfer
Catalog # EHERCA11433000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pk:25 mg
- Standards:Reference standard
- CAS No.:26172-55-4
- Description:5-Chloro-2-methyl-4-isothiazolin-3-one
- Formula:C₄H₄ClNOS
- Description:5-Chloro-2-methyl-4-isothiazolin-3-one
Specifications
About this item
This reference material is used as a calibrant for chromatography or any other analytical technique.