Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Virginiamycin complex ≥95%

Virginiamycin complex ≥95%

Supplier:   Cayman Chemical
Virginiamycin complex ≥95%
Supplier:  Cayman Chemical
Certificates
Virginiamycin complex ≥95%

3 Options Available Below

Product Details & Documents

Virginiamycin complex is a complex containing two streptogramin antibiotics, virginiamycin M1 (75%) and virginiamycin S1 (25%), produced by S. virginiae.

As a complex, the two antibiotics act synergistically to irreversibly inhibit protein synthesis in bacteria. When given to livestock, virginiamycin improves growth. Virginiamycin complex is also used in fuel ethanol fermenters to prevent to control bacteria competing with yeast for nutrients.

  • CAS Number:121006-76-1
  • Storage Temperature:Freezer
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 110,00 €
    110,00 €
    1 * 1 mg
       110,00 €

  • 5 mg
    Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 486,00 €
    486,00 €
    1 * 5 mg
       486,00 €

  • 10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 859,00 €
    859,00 €
    1 * 10 mg
       859,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.