Order Entry
Netherlands
ContactUsLinkComponent
Lithium tetrakis(pentafluorophenyl)borate ethyl etherate
Irritant
Lithium tetrakis(pentafluorophenyl)borate ethyl etherate
Certificates
undefined
Irritant
Lithium tetrakis(pentafluorophenyl)borate ethyl etherate

Specification Test Results

  • Appearance:
    White to light beige Powder or crystals
  • Melting point:
    117 to 122°C
  • NMR (19F-NMR):
    ≥96.0 %

Specification Test Results

Frequently Bought Together