Order Entry
Netherlands
ContactUsLinkComponent
Ciprofloxacin hydrochloride hydrate 98%
Ciprofloxacin hydrochloride hydrate 98%
Certificates
undefined
Ciprofloxacin hydrochloride hydrate 98%

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    4.0 to 7.0 %
  • HPLC:
    ≥97.5 %
  • Residue after ignition:
    ≤0.1 %

Specification Test Results

Frequently Bought Together