Order Entry
Netherlands
ContactUsLinkComponent
 
Irritant
Minocycline hydrochloride 97%
Catalog # ACRO455330010
CAS Number:  
undefined
Irritant
Minocycline hydrochloride 97%
Catalog # ACRO455330010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow to ochre Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤10 %
  • HPLC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :493.94 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:13614-98-7
Formula:C₂₃H₂₇N₃O₇
MDL Number:MFCD00083669