(+)-cis-Diltiazem hydrochloride 98%
: (2S,3S)-(+)-3-Acetoxy-2,3-dihydro-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one Hydrochloride, Diltiazem Hydrochloride, (+)-cis-Diltiazem HCl, (+)-cis-3-Acetoxy-5-(2-dimethylaminoethyl)-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one Hydrochloride, (2S,3S)-5-(2-(Dimethylamino)ethyl)-2-(4-methoxyphenyl)-4-oxo-2,3,4,5-tetrahydrobenzo[b][1,4]thiazepin-3-yl acetate hydrochloride
:33286-22-5
:
:C₂₂H₂₆N₂O₄S·HCl
:MFCD00069252
:450.98 g/mol
:210…215 °C
:GHS07
:Refrigerator
- Environmentally Preferable:
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Specific optical rotation:+110° to +120° (20°C, 589 nm) (c=1, H2O)
Frequently Bought Together