Order Entry
Netherlands
ContactUsLinkComponent
 
Flunixin Related Compounds, Reference Standard
 :  USP
 
Flunixin Related Compounds, Reference Standard
 :  USP

 

  • CAS No.:
    54396-44-0
  • UN No.:
    2811
  • Synonym(s):
    2-Methyl-3-(trifluoromethyl)aniline
  • Formula:
    C₈H₈F₃N
  • Description:
    Flunixin Related Compound B

 

Frequently Bought Together