Order Entry
Netherlands
ContactUsLinkComponent
Organic reference standard, citric acid, CRM, TraceCERT®, Supelco®
Organic reference standard, citric acid, CRM, TraceCERT®, Supelco®
  94676-100MG
 :  Merck
 :  
undefined
Organic reference standard, citric acid, CRM, TraceCERT®, Supelco®
  94676-100MG
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Standards:
    Reference standard
  • CAS No.:
    77-92-9
  • Description:
    Citric acid
  • Formula:
    C₆H₈O₇
  • Description:
    Citric acid