Order Entry
Netherlands
ContactUsLinkComponent
 
 
Nimodipine
 
undefined
 
Nimodipine

 

  • Appearance:
    Yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    124 to 130 °C
  • Loss on drying:
    ≤0.5 % (105 °C, 4 h)
  • HPLC:
    ≥97.5 %

 

Frequently Bought Together