Organic reference standards, cyclazadone (NMIA D914), Certified Reference Material (CRM)
Catalog # PROONMIAD914
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:10 mg
- Standards:Reference standard
- CAS No.:14461-91-7
- Description:Cyclazadone
- Environmentally Preferable:
- Formula:C₁₂H₁₂N₂O₂
- Description:Cyclazadone
Specifications
About this item
The purity value obtained from a combination of traditional analytical techniques is 99,6 ± 1,4% mass fraction, (95% coverage interval). The purity estimate by traditional analytical techniques was obtained by subtraction from 100% of total impurities by GC-FID, thermogravimetric analysis, Karl Fischer analysis and 1H NMR and also supported by elemental microanalysis.
- White solid
- Moisture content ≤0,3% mass fraction
- Volatile residue ≤0,1% mass fraction
- Non volatile residue ≤0,2% mass fraction
- CAS no.: 14461-91-7
Synonyms: 5-phenyl-2-cyclopropylamino-4-oxazolinone; 4(5H)-oxazolone-2-(cyclopropylamino)-5-phenyl-cyclopropylpemoline.