Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Methyldopa, Reference Standards

Methyldopa, Reference Standards

Supplier:   USP
Methyldopa, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1426002.
  • 1427027.
  • 1427038.
  • Description:
  • Methyldopa
  • Methyldopa Related Compound B
  • Methyldopa Related Compound C
  • Synonym(s):
  • (S)-2-Amino-3-(4-methoxyphenyl)-2-methylpropanoic acid hydrochloride
  • (S)-2-Amino-3-(3,4-dimethoxyphenyl)-2-methylpropanoic acid hydrochloride monohydrate
  • CAS No.:
  • 41372-08-1
  • 35026-10-9
  • Formula:
  • C₁₀H₁₅NO₅

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1426002.
    Methyldopa
    Methyldopa
    USP reference standard
    Vial
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 688,00 €
    688,00 €
    1 * 500 mg
       688,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1426002.
    Description
    Methyldopa
    Synonym(s)
    CAS No.
    41372-08-1
    Formula
    C₁₀H₁₅NO₅
  • 1427027.
    Methyldopa Related Compound B
    Methyldopa Related Compound B
    USP reference standard
    Vial
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 1 270,00 €
    1 270,00 €
    1 * 10 mg
       1 270,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1427027.
    Description
    Methyldopa Related Compound B
    Synonym(s)
    (S)-2-Amino-3-(4-methoxyphenyl)-2-methylpropanoic acid hydrochloride
    CAS No.
    35026-10-9
    Formula
  • 1427038.
    Methyldopa Related Compound C
    Methyldopa Related Compound C
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 1 270,00 €
    1 270,00 €
    1 * 20 mg
       1 270,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1427038.
    Description
    Methyldopa Related Compound C
    Synonym(s)
    (S)-2-Amino-3-(3,4-dimethoxyphenyl)-2-methylpropanoic acid hydrochloride monohydrate
    CAS No.
    Formula
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.