Afterwash solution
Afterwash solution, 200 ml bottle
Catalog # LBES8205212
Supplier: LABORATOIRES ESCULAPE
Jump to:
Product Details & Documents
This afterwash solution quickly restores the natural physiological condition of the eye following decontamination with Diphoterine® or Hexafluorine® solutions.
- Ergonomically eye-shaped cup ensures a tighter seal around the eye to maximise efficiency during rinsing
- Helps to restore the physiological state of the cornea more quickly and more smoothly
- May be used for eye wash in case of non chemical solid particles
Shelf life: 2 years
Specifications
- Description:Afterwash solution, 200 ml bottle
- Seq:03