Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Lamotrigine, Reference Standards

Lamotrigine, Reference Standards

Supplier:   USP
Lamotrigine, Reference Standards
Supplier:  USP
Certificates

4 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1356756.
  • 1356778.
  • 1356789.
  • 1356790.
  • Description:
  • Lamotrigine
  • Lamotrigine Related Compound B
  • Lamotrigine Related Compound C
  • Lamotrigine Related Compound D
  • Synonym(s):
  • 2,3-dichlorobenzoic acid
  • 3-amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-5(4H)-one
  • N-[5-amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-3-yl]-2,3-dichlorobenzamide
  • CAS No.:
  • 84057-84-1
  • 50-45-3
  • 252186-78-0
  • 252186-79-1
  • Formula:
  • C₉H₇Cl₂N₅
  • C₇H₄Cl₂O₂
  • C₉H₆Cl₂N₄O
  • C₁₆H₉Cl₄N₅O
  • UN No.:
  • 2811

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1356756.
    Lamotrigine
    Lamotrigine
    USP reference standard
    Vial
    150 mg
    Log in for availability
    Log in for availability
    1 * 150 mg
    1 * 150 mg - 1 010,00 €
    1 010,00 €
    1 * 150 mg
       1 010,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1356756.
    Description
    Lamotrigine
    Synonym(s)
    CAS No.
    84057-84-1
    Formula
    C₉H₇Cl₂N₅
    UN No.
    2811
  • 1356778.
    Lamotrigine Related Compound B
    Lamotrigine Related Compound B
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 1 240,00 €
    1 240,00 €
    1 * 20 mg
       1 240,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1356778.
    Description
    Lamotrigine Related Compound B
    Synonym(s)
    2,3-dichlorobenzoic acid
    CAS No.
    50-45-3
    Formula
    C₇H₄Cl₂O₂
    UN No.
  • 1356789.
    Lamotrigine Related Compound C
    Lamotrigine Related Compound C
    USP reference standard
    Vial
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 1 260,00 €
    1 260,00 €
    1 * 10 mg
       1 260,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1356789.
    Description
    Lamotrigine Related Compound C
    Synonym(s)
    3-amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-5(4H)-one
    CAS No.
    252186-78-0
    Formula
    C₉H₆Cl₂N₄O
    UN No.
  • 1356790.
    Lamotrigine Related Compound D
    Lamotrigine Related Compound D
    USP reference standard
    Vial
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 1 150,00 €
    1 150,00 €
    1 * 10 mg
       1 150,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1356790.
    Description
    Lamotrigine Related Compound D
    Synonym(s)
    N-[5-amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-3-yl]-2,3-dichlorobenzamide
    CAS No.
    252186-79-1
    Formula
    C₁₆H₉Cl₄N₅O
    UN No.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.