Order Entry
Netherlands
ContactUsLinkComponent
Fluvastatin Related Compounds, Reference Standard
Fluvastatin Related Compounds, Reference Standard
Supplier:  USP
Certificates
Fluvastatin Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    93957-55-2
  • Synonym(s):
    Fluvastatin sodium and fluvastatin sodium anti-isomer ([R*,R*-E]-(+/-)-7-[3-(4-Fluorophenyl)-1-(methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-6-heptenoic acid monosodium salt)
  • Formula:
    C₂₄H₂₅FNNaO₄
  • Description:
    Fluvastatin for

Specifications

Frequently Bought Together