Human recombinant Flagellin (from Salmonella typhimurium strain 14028)
Catalog # PRSI90-236
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:Salmonella typhimurium strain 14028
- Species:Human
- Storage conditions:Stable for at least 6 months after receipt when stored at −20 °C.
- Biological activity:Activation of TLR5 in human epithelial cell assays based on NF-kappaB luciferase fusions, on expression of IL-8 (CXCL8), CCL20, inducible nitric oxide synthase, beta-defensin 2 and matrilysin (MMP-7). Maturation of dendritic cells in in vitro and in vivo assays. Adjuvant activity on foreign antigens delivered by subcutaneous and intranasal routes in mice. TLR5 can be stimulated in vitro by Flagellin (high Purity) at a concentration of 5-50ng /ml and in vivo at a starting concentration of 0.1-1ug /mouse.
- Protein synonyms:FliC
- Protein/peptide name:Flagellin
- Purity:>95% (SDS-PAGE)
- Formulation:Lyophilized, Contains PBS.
- Tested applications:Research use only
Specifications
About this item
Flagellin is the subunit protein which polymerises to form the filaments of bacterial flagella. It activates the innate immune system through the receptor Toll-like Receptor 5 (TLR5) and the intracellular protein NLRC4 (NLR family CARD domain-containing protein 4).
Fusion-Tag:
This recombinant proteins is for research use only.