Order Entry
Netherlands
ContactUsLinkComponent
Human recombinant IL35 (from cells)
Human recombinant IL35 (from cells)
Catalog # PRSI92-680
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IL35 (from cells)
Catalog # PRSI92-680
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Interleukin 35|IL35|hIL35
  • Protein/peptide name:
    IL35
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    73.9 kD
  • Sequence:
    Arg21-Lys229&Arg23-Ser219
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Interleukin-35 is a newest and potent suppressive cytokine of the IL-12 family. Unlike other members of the IL-12 family which are produced mainly by antigen-presenting cells (APCs), IL-35 has been identified as a potentially immunosuppressive cytokine primarily produced by regulatory T (Treg) cells, and to a lesser extent by activated dendritic cells and macrophages. IL-35 is composed of the IL-12 p35 subunit and the IL-27 Epstein–Barr virus-induced protein 3 (EBI3) subunit. IL-35 plays an important role in many disease models and has been recently shown to have additional functions aside from inhibition of proliferation, including inducing its own expression in non-Treg cells.

Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.