Order Entry
Netherlands
ContactUsLinkComponent
Human recombinant Irisin (from cells)
Human recombinant Irisin (from cells)
  PRSI92-628
 :  
Human recombinant Irisin (from cells)
  PRSI92-628
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Irisin|Fibronectin type III repeat-containing protein 2|Fibronectin type III domain-containing protein 5|FNDC5
  • Protein/peptide name:
    Irisin
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    39.7 kD
  • Sequence:
    Asp32-Glu143
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

 

 

FNDC5 is a membrane protein comprised by a short cytoplasmic domain, a transmembrane segment, and an ectodomain consisting of a fibronectin type III (FNIII) domain. The ectodomain has been proposed to be cleaved to a soluble peptide hormone named “irisin”. Irisin is secreted from muscle in response to exercise, and may regulate energy metabolism, stem cell differentiation, and neuronal development. The circulating irisin converts white fat into the more thermogenic beige fat, and may mediate some beneficial effects of exercise in humans and the potential for generating weight loss. In mice, expression of Irisin has been shown to regulate obesity and diabetes. A similar function is suggested in human.

Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.