Order Entry
Netherlands
ContactUsLinkComponent
Mouse recombinant NGR
Mouse recombinant NGR
Catalog # PRSI92-608
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant NGR
Catalog # PRSI92-608
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    NgR|NOGOR|Nogo Receptor|RTN4R|Nogo-66 Receptor|Reticulon-4 Receptor
  • Protein/peptide name:
    NGR
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    72.7 kD
  • Sequence:
    Cys27-Ser447
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

Nogo Receptor (NgR) is a glycosylphosphoinositol (GPI)-anchored protein that belongs to the Nogo receptor family. Human NgR is predominantly expressed in neurons and their axons in the central nervous systems. As a receptor for myelin-derived proteins Nogo, myelin-associated glycoprotein (MAG) and myelin oligodendrocyte glycoprotein (OMG), NgR mediates axonal growth inhibition and may play a role in regulating axonal regeneration and plasticity in the adult central nervous system. NgR may be proposed as a potential drug target for treatment of various neurological conditions. Additionally, NgR may play a role in regulating the function of gap junctions.

Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.