Order Entry
Netherlands
ContactUsLinkComponent
 
Mouse recombinant IL-12 R beta 2
Catalog # PRSI92-584
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant IL-12 R beta 2
Catalog # PRSI92-584
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    IL-12RB2|interleukin-12 receptor subunit beta-2|IL-12R subunit beta-2|IL-12 receptor beta 2|IL12RB2|interleukin-12 receptor beta-2 chain|IL-12R-beta-2|IL-12 receptor subunit beta-2
  • Protein/peptide name:
    IL-12 R beta 2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    98.4 kD
  • Sequence:
    Met1-Asn637
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

The IL12 receptor complex, formed by IL12RB1 and IL12RB2, mediates the type I immune responses of various types of lymphocytes. Its ligand, IL12, is a heterodimeric cytokine composed of IL-12p35 and IL-12p40 subunits that are linked via disulphide bonds. Ligation of IL-12 to its receptor involves the binding of IL-12p35 to IL12RB1 and IL-12p40 to IL12RB2. This will result in the activation of tyrosine kinase 2 (TYK2), which is associated with the IL12RB1 chain and Janus kinase 2 (JAK2), which is associated with the IL12RB2 chain. Activated TYK2 and JAK2 direct the phosphorylation of STAT4. IL12RB1 is present on all lymphocytes, while the expression of IL12RB2 is tightly regulated. It has shown that the expression of IL12RB2 is limited to Th2 cells. IL12RB2 subunit plays an important role in Th1 cell differentiation, since its absence leads to an abortive Th1 differentiation that has dysfunctional production of Th1 effector molecules.

Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.