Order Entry
Netherlands
ContactUsLinkComponent
Human recombinant Thymic stromal lymphopoietin (from E. coli)
Human recombinant Thymic stromal lymphopoietin (from E. coli)
Catalog # PRSI92-513
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Thymic stromal lymphopoietin (from E. coli)
Catalog # PRSI92-513
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Thymic stromal lymphopoietin/TSLP|TSLP
  • Protein/peptide name:
    Thymic stromal lymphopoietin
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    15.1 kD
  • Sequence:
    Tyr29-Gln159
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

Thymic stromal lymphopoietin (TSLP) is a novel member of the hemopoietic cytokine family that promotes the development of B cells and shares overlapping activity with IL-7. The human TSLP protein comprises a 28 amino acids (aa) signal sequence and 131 aa mature region. Human TSLP has two isoforms lfTSLP (159 aa) and sfTSLP (63 aa) produced by alternative splicing . lfTSLP (159aa) is expressed in a number of tissues including heart, liver and prostate, and sfTSLP (63aa) is predominantly expressed in keratinocytes of oral mucosa, skin and in salivary glands. In aa sequence level, Human TSLP displays about 43% identity with mouse TSLP. Thymic stromal lymphopoietin (TSLP) is a cytokine that functions mainly on myeloid cells; it induces the release of T cell-attracting chemokines from monocytes and enhances the maturation of CD11c(+) dendritic cells . TSLP has proliferative effects on the myeloid cell line and may initiate asthma or atopic dermatitis responses by directly activating mast cells . TSLP signals cells via the interleukin-7 receptor- alpha chain (IL-7R alpha), shared with IL-7, together with the TSLP receptor (TSLPR) subunit. Recent studies indicate that TSLP and its receptor are novel therapeutic targets for rheumatoid arthritis , for increased intraarticular TSLP concentrations in patients has caused chemotaxis and activation of arthritogenic T cells.

This recombinant protein can be used for biological assays. For research use only.