Order Entry
Netherlands
ContactUsLinkComponent
Mouse recombinant Reg2
Mouse recombinant Reg2
Catalog # PRSI92-438
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant Reg2
Catalog # PRSI92-438
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Regenerating Islet-derived 2|PSP|Islet of Langerhans regenerating protein 2|PTP|Lithostathine-2|Pancreatic stone protein 2|Pancreatic thread protein 2
  • Protein/peptide name:
    Reg2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    17.7 kD
  • Sequence:
    Gln23-Ala173
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Regenerating protein 2 (Reg2) also known as Lithostathine 2, pancreatic thread protein (PTP2) and pancreatic stone protein 2 (PSP2), is a member of the Reg family of proteins. These small, secreted proteins have been implicated in a range of physiological processes including acting as acute phase reactants, lectins, survival/growth factors for insulin-producing pancreatic beta-cells, neural cells, and epithelial cells of the digestive system. Studies also indicate a role for Reg family members in tumour formation and indicate their potential for use as biomarkers of carcinogenesis. Mouse Reg2 is expressed in regenerating islets and normal exocrine pancreas. Reg2 also stimulates the growth of pancreatic beta cells. Mouse Reg2 belongs to the type II subclass of the Reg family and is the only subclass II Reg protein described.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.