Order Entry
Netherlands
ContactUsLinkComponent
Human recombinant Yin and Yang 1 (from E. coli)
Human recombinant Yin and Yang 1 (from E. coli)
Catalog # PRSI92-275
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Yin and Yang 1 (from E. coli)
Catalog # PRSI92-275
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Delta transcription factor|INO80S|YY1|Transcriptional repressor protein YY1|NF-E1|Yin and yang 1|INO80 complex subunit S
  • Protein/peptide name:
    Yin and Yang 1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    12.6 kD
  • Sequence:
    Val221-Gly321
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Transcriptional repressor protein YY1(YY1)contains 4 C2H2-type zinc fingers and belongs to the YY transcription factor family. Multifunctional transcription factor exhibits positive and negative control on a large number of cellular and viral genes by binding to sites overlapping the transcription start site. The effect on transcription regulation of the protein is depending upon the context in which it binds and diverse mechanisms of action include direct activation or repression, indirect activation or repression via cofactor recruitment, or activation or repression by disruption of binding sites or conformational DNA changes. Its activity is regulated by transcription factors and cytoplasmic proteins that have been shown to abrogate or completely inhibit YY1-mediated activation or repression.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.