Order Entry
Netherlands
ContactUsLinkComponent
Human recombinant PSD95 (from E. coli)
Human recombinant PSD95 (from E. coli)
Catalog # PRSI92-242
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant PSD95 (from E. coli)
Catalog # PRSI92-242
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Disks large homolog 4|SAP-90|SAP90|DLG4|PSD-95|Postsynaptic density protein 95|Synapse-associated protein 90|PSD95
  • Protein/peptide name:
    PSD95
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    81.9 kD
  • Sequence:
    Met1-Leu724
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Disks large homolog 4(DLG4) is a cell membrane protein and it is a member of the membrane-associated guanylate kinase (MAGUK) family. The protein contains 1 guanylate kinase-like domain,3 PDZ (DHR) domains and 1 SH3 domain. With PSD-93 it is recruited into the same NMDA receptor and potassium channel clusters. These two MAGUK proteins may interact at postsynaptic sites to form a multimeric scaffold for the clustering of receptors, ion channels, and associated signaling proteins. DLG4 is the best studied member of the MAGUK-family of PDZ domain-containing proteins. Like all MAGUK-family proteins, its basic structure includes three PDZ domains, an SH3 domain, and a guanylate kinase-like domain (GK) connected by disordered linker regions. It is almost exclusively located in the post synaptic density of neurons, and is involved in anchoring synaptic proteins. Its direct and indirect binding partners include neuroligin, NMDA receptors, AMPA receptors, and potassium channels.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.